Product Details

SNP ID
rs35636070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16124349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGTGTCTGTCTTCACACTCAC[A/G]GCGCTCTCGGCAGACAGGTAAGTAC
Phenotype
MIM: 305670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRPR PubMed Links

Gene Details

Gene
GRPR
Gene Name
gastrin releasing peptide receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005314.2 1049 Silent Mutation ACA,ACG T132T NP_005305.1

View Full Product Details