Product Details

SNP ID
rs17049006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:117085151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCAGCATGTAGAAGAAAACCGT[A/G]GCCAAATCTTTGACACCATAATAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAM1L1 PubMed Links

Gene Details

Gene
TRAM1L1
Gene Name
translocation associated membrane protein 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152402.2 430 Silent Mutation GCC,GCT A81A NP_689615.2

View Full Product Details