Product Details

SNP ID
hCV25638143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140786283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAGCATAAGAAAGGTGTAGTC[C/T]TTTTGCAATGGTGTTTTCTAGGAGA
Phenotype
MIM: 606307 MIM: 606308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018900.3 148 UTR 5 NP_061723.1
NM_031410.2 148 UTR 5 NP_113598.1
NM_031411.2 148 UTR 5 NP_113599.1
Gene
PCDHA2
Gene Name
protocadherin alpha 2
There are no transcripts associated with this gene.

View Full Product Details