Product Details

SNP ID
rs61743807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116790229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCAGCTTGCTCAGAACCTTGC[C/T]ACTGTCTGTTTGTTGAAACTCTGGG
Phenotype
MIM: 606368 MIM: 603901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOA5 PubMed Links

Gene Details

Gene
APOA5
Gene Name
apolipoprotein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166598.1 1073 Missense Mutation AGC,GGC S334G NP_001160070.1
NM_052968.4 1073 Missense Mutation AGC,GGC S334G NP_443200.2
XM_017017169.1 1073 Missense Mutation AGC,GGC S334G XP_016872658.1
Gene
ZPR1
Gene Name
ZPR1 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317086.1 1073 Intron NP_001304015.1
NM_003904.4 1073 Intron NP_003895.1

View Full Product Details