Product Details

SNP ID
rs61745027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102937393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCTGGTGGGCGACAGGGACGT[A/G]GGCAAGAGTGAGATCCTGGAGAGCC
Phenotype
MIM: 300405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB40AL PubMed Links

Gene Details

Gene
RAB40AL
Gene Name
RAB40A, member RAS oncogene family-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031834.1 122 Silent Mutation GTA,GTG V25V NP_001027004.1

View Full Product Details