Product Details

SNP ID
rs35945487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50907030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTTGGCCACACGGGGCTTTTCC[A/G]AAAGACACAAGGCCCTGCAAGTACC
Phenotype
MIM: 603767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK4 PubMed Links

Gene Details

Gene
KLK4
Gene Name
kallikrein related peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302961.1 669 Silent Mutation TTC,TTT F128F NP_001289890.1
NM_004917.4 669 Silent Mutation TTC,TTT F223F NP_004908.4
XM_005259441.4 669 Silent Mutation TTC,TTT F128F XP_005259498.2
XM_011527545.2 669 Intron XP_011525847.1
XM_011527546.2 669 UTR 3 XP_011525848.1

View Full Product Details