Product Details

SNP ID
rs34771052
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:204618017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGAGAGGCCGCCTCCCACCC[A/G]CACCCTTCCTGGGTTGGCCTGTGCC
Phenotype
MIM: 605492
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371692 PubMed Links

Gene Details

Gene
LOC105371692
Gene Name
uncharacterized LOC105371692
There are no transcripts associated with this gene.

Gene
LRRN2
Gene Name
leucine rich repeat neuronal 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006338.2 2434 Missense Mutation GCG,GTG A659V NP_006329.2
NM_201630.1 2434 Missense Mutation GCG,GTG A659V NP_963924.1
XM_005244827.3 2434 Missense Mutation GCG,GTG A659V XP_005244884.1
XM_011509066.2 2434 Missense Mutation GCG,GTG A659V XP_011507368.1
XM_017000050.1 2434 Missense Mutation GCG,GTG A659V XP_016855539.1

View Full Product Details