Product Details

SNP ID
rs140013528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37591047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAGCTGGTGTCATGCTGAGCAT[A/G]CAGATGCATTTGCTCCCTGGATGCA
Phenotype
MIM: 611212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf19 PubMed Links

Gene Details

Gene
C4orf19
Gene Name
chromosome 4 open reading frame 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104629.1 1239 UTR 3 NP_001098099.1
NM_018302.2 1239 UTR 3 NP_060772.2
XM_011513712.2 1239 UTR 3 XP_011512014.1
XM_011513713.2 1239 UTR 3 XP_011512015.1
XM_017008361.1 1239 UTR 3 XP_016863850.1
Gene
RELL1
Gene Name
RELT like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085399.1 1239 UTR 3 NP_001078868.1
NM_001085400.1 1239 Intron NP_001078869.1
XM_017008590.1 1239 Intron XP_016864079.1

View Full Product Details