Product Details

SNP ID
rs114740909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52586228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCAGCTCTAGCCCCACGTCAGAG[C/T]CCCTTCTTGGGCATCTCGGGCCACC
Phenotype
MIM: 608246
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT72 PubMed Links

Gene Details

Gene
KRT72
Gene Name
keratin 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146225.1 Intron NP_001139697.1
NM_001146226.1 Intron NP_001139698.1
NM_080747.2 Intron NP_542785.1
XM_017018833.1 Intron XP_016874322.1
XM_017018834.1 Intron XP_016874323.1
XM_017018835.1 Intron XP_016874324.1
XM_017018836.1 Intron XP_016874325.1
XM_017018837.1 Intron XP_016874326.1

View Full Product Details