Product Details
- SNP ID
-
hCV25638817
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:70259687 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTTACAGAGACGTTGGTAAAACT[C/G]TTTTCCTCACTTTCTTGTTAGTTGG
- Phenotype
-
MIM: 610995
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1285-2
PubMed Links
Gene Details
- Gene
- MIR1285-2
- Gene Name
- microRNA 1285-2
There are no transcripts associated with this gene.
- Gene
- PCYOX1
- Gene Name
- prenylcysteine oxidase 1
There are no transcripts associated with this gene.
View Full Product Details