Product Details

SNP ID
rs114934326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5455418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCATCTAGAACCCTGGTGACTCC[C/T]GGTCTCCATGTCATCTGCCAGAAAG
Phenotype
MIM: 612063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNRF4 PubMed Links

Gene Details

Gene
ZNRF4
Gene Name
zinc and ring finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181710.3 4 UTR 5 NP_859061.3

View Full Product Details