Product Details
- SNP ID
-
rs3795262
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
60
- Location
-
Chr.1:3022133 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- AGGCGGTGCTGGCTCTCTACGCCTC[G/T]GCCTGTGTCACGGGCCTGGTGGTGG
- Phenotype
-
MIM: 608535
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ACTRT2
PubMed Links
Gene Details
- Gene
- ACTRT2
- Gene Name
- actin related protein T2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080431.4 |
652 |
Silent Mutation |
TCG,TCT |
S149S |
NP_536356.3 |
View Full Product Details