Product Details

SNP ID
rs34654901
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:159065976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGTTTGAAGCGTGTTGATCTT[A/C]GGGGTTTCACCAGCTTTTCTGATAA
Phenotype
MIM: 604578
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AIM2 PubMed Links

Gene Details

Gene
AIM2
Gene Name
absent in melanoma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004833.1 995 Silent Mutation NP_004824.1
XM_005245616.4 995 Silent Mutation XP_005245673.1
XM_017002848.1 995 Silent Mutation XP_016858337.1

View Full Product Details