Product Details

SNP ID
rs61734000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27890414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCGCCACGGATACGACGCGCAA[A/G]CTGGATGTCCTTAGGCATAATAGTG
Phenotype
MIM: 602796 MIM: 602808 MIM: 602817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AM PubMed Links

Gene Details

Gene
HIST1H2AM
Gene Name
histone cluster 1, H2am
There are no transcripts associated with this gene.

Gene
HIST1H2BO
Gene Name
histone cluster 1, H2bo
There are no transcripts associated with this gene.

Gene
HIST1H3J
Gene Name
histone cluster 1, H3j
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003535.2 Intron NP_003526.1

View Full Product Details