Product Details

SNP ID
rs61734022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46574447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTCCGTGCTGCGGCTGTACGC[C/G]CCGCCGGGGGACCCCGCTGCGCGCG
Phenotype
MIM: 300375
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHST7 PubMed Links

Gene Details

Gene
CHST7
Gene Name
carbohydrate sulfotransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019886.3 761 Silent Mutation GCC,GCG A172A NP_063939.2

View Full Product Details