Product Details

SNP ID
rs117159068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3440454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTCAAAATGCTATAATTCCTT[C/T]ACGTGAGTCAGGGCCCCACTGCTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1E2 PubMed Links

Gene Details

Gene
OR1E2
Gene Name
olfactory receptor family 1 subfamily E member 2
There are no transcripts associated with this gene.

Gene
SPATA22
Gene Name
spermatogenesis associated 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170695.1 Intron NP_001164166.1
NM_001170696.1 Intron NP_001164167.1
NM_001170697.1 Intron NP_001164168.1
NM_001170698.1 Intron NP_001164169.1
NM_001170699.1 Intron NP_001164170.1
NM_001321336.1 Intron NP_001308265.1
NM_001321337.1 Intron NP_001308266.1
NM_032598.4 Intron NP_115987.2

View Full Product Details