Product Details

SNP ID
rs34433477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70335843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGAGAAGTTTATCACTGTGTT[C/T]GAACTCACTACCTCTGCTGAAAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABS1 PubMed Links

Gene Details

Gene
CABS1
Gene Name
calcium binding protein, spermatid associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033122.3 890 Silent Mutation TTC,TTT F268F NP_149113.3

View Full Product Details