Product Details

SNP ID
rs3752418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26045509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCGTAAGAGCGCTCCGGCCAC[G/C]GGTGGCGTGAAGAAACCTCATCGCT
Phenotype
MIM: 602803 MIM: 602812
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2BB PubMed Links

Gene Details

Gene
HIST1H2BB
Gene Name
histone cluster 1, H2bb
There are no transcripts associated with this gene.

Gene
HIST1H3C
Gene Name
histone cluster 1, H3c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003531.2 Intron NP_003522.1

View Full Product Details