Product Details

SNP ID
rs17184009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29440178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTCTCCACTGATGCTGCCCTC[C/T]AGTCCCCTATGTACTTCTTCCTGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10C1 PubMed Links

Gene Details

Gene
OR10C1
Gene Name
olfactory receptor family 10 subfamily C member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013941.3 240 Nonsense Mutation CAG,TAG Q55* NP_039229.3
Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 240 Intron NP_110503.3
XM_011514929.2 240 Intron XP_011513231.1
XM_011514930.2 240 Intron XP_011513232.1

View Full Product Details