Product Details

SNP ID
rs80327720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131260807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTATTTGTGAGTCTAACCTAGC[A/G]GGTGGTCCTGGCTGTCACCGGTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM78A PubMed Links

Gene Details

Gene
FAM78A
Gene Name
family with sequence similarity 78 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033387.3 1207 UTR 3 NP_203745.2
XM_011518567.1 1207 UTR 3 XP_011516869.1
XM_011518568.2 1207 UTR 3 XP_011516870.1

View Full Product Details