Product Details

SNP ID
rs28730867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169509786 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCAGGTTTCTCACAGAATGGATT[C/T]CCAGCGGGAACTTGCAGAGGAACTG
Phenotype
MIM: 603650 MIM: 607701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BBS5 PubMed Links

Gene Details

Gene
BBS5
Gene Name
Bardet-Biedl syndrome 5
There are no transcripts associated with this gene.

Gene
KLHL41
Gene Name
kelch like family member 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006063.2 85 Missense Mutation TCC,TTC S3F NP_006054.2

View Full Product Details