Product Details
- SNP ID
-
rs3732388
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:192798641 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATGGCGGTGCGCAAGTGCTCTGC[C/T]GGGTCTGAGCGCACAGAGGACAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MB21D2
PubMed Links
Gene Details
- Gene
- MB21D2
- Gene Name
- Mab-21 domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178496.3 |
1542 |
Silent Mutation |
CCA,CCG |
P407P |
NP_848591.2 |
View Full Product Details