Product Details

SNP ID
rs73546949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58001433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCGGCACAGCGGAACTCCGGGT[C/G]CCGGTTGAGGTTGCTGGTGGACCTG
Phenotype
MIM: 613276
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USB1 PubMed Links

Gene Details

Gene
USB1
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195302.1 27 UTR 5 NP_001182231.1
NM_001204911.1 27 UTR 5 NP_001191840.1
NM_024598.3 27 UTR 5 NP_078874.2
XM_005256144.3 27 Intron XP_005256201.1
XM_011523328.2 27 UTR 5 XP_011521630.1
XM_011523329.2 27 Intron XP_011521631.1
XM_011523330.1 27 UTR 5 XP_011521632.1
Gene
ZNF319
Gene Name
zinc finger protein 319
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020807.2 27 Intron NP_065858.1
XM_005256069.3 27 Intron XP_005256126.1

View Full Product Details