Product Details

SNP ID
rs55969078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56113203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGATGGCAAAGGAATAAATGTC[G/T]GCTTTAGGCGTCACGCCCTCTCCTT
Phenotype
MIM: 190060
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOS PubMed Links

Gene Details

Gene
MOS
Gene Name
v-mos Moloney murine sarcoma viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005372.1 780 Silent Mutation NP_005363.1

View Full Product Details