Product Details

SNP ID
rs545207483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30164252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGTGCTTTACAATTAGATACC[A/G]TCTTCCATGTTTATTTCACTGATGT
Phenotype
MIM: 612963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN6 PubMed Links

Gene Details

Gene
DCTN6
Gene Name
dynactin subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006571.3 Intron NP_006562.1

View Full Product Details