Product Details

SNP ID
rs147451538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111262327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAACGTGAACCGAAATATCAGTA[C/G]TCTGTCTTTCATTTTTGTGGGTCGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MFSD4B PubMed Links

Gene Details

Gene
MFSD4B
Gene Name
major facilitator superfamily domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153369.2 520 Missense Mutation ACT,AGT T33S NP_699200.2
XM_017011483.1 520 Missense Mutation ACT,AGT T33S XP_016866972.1

View Full Product Details