Product Details

SNP ID
hCV25642967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:243270156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACATGTGCCCTGAAAGAAGGCG[A/T]TGTCACTATTGGAGAAGATGCACCA
Phenotype
MIM: 613524
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SDCCAG8 PubMed Links

Gene Details

Gene
SDCCAG8
Gene Name
serologically defined colon cancer antigen 8
There are no transcripts associated with this gene.

View Full Product Details