Product Details

SNP ID
rs543453041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176596418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACTTGTATGCAGTCTCTCCCTG[T/A]TCTGGGGATCTGGCCCCACCGGAGA
Phenotype
MIM: 611239
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
CDHR2 PubMed Links

Gene Details

Gene
CDHR2
Gene Name
cadherin related family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171976.1 3592 Intron NP_001165447.1
NM_017675.4 3592 Intron NP_060145.3
Gene
GPRIN1
Gene Name
G protein regulated inducer of neurite outgrowth 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052899.2 3592 UTR 3 NP_443131.2

View Full Product Details