Product Details

SNP ID
rs143574108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185403398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCCACACAGGGAGATCTCTGC[C/T]CTCAGCTTATTGTCTTTCTGTTACC
Phenotype
MIM: 611482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD37 PubMed Links

Gene Details

Gene
ANKRD37
Gene Name
ankyrin repeat domain 37
There are no transcripts associated with this gene.

Gene
C4orf47
Gene Name
chromosome 4 open reading frame 47
There are no transcripts associated with this gene.

Gene
LRP2BP
Gene Name
LRP2 binding protein
There are no transcripts associated with this gene.

Gene
UFSP2
Gene Name
UFM1 specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018359.3 Intron NP_060829.2

View Full Product Details