Product Details

SNP ID
rs12827843
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:122204614 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAGCACGGAGACAGTCTCCAGCTGC[C/G]GTTCATGCTTCCTCCCCAGCCTTCC
Phenotype
MIM: 605864 MIM: 605219
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
B3GNT4 PubMed Links
Additional Information
For this assay, SNP(s) [rs145969982] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
B3GNT4
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538732.1 Intron XP_011537034.1
Gene
DIABLO
Gene Name
diablo IAP-binding mitochondrial protein
There are no transcripts associated with this gene.

Gene
LRRC43
Gene Name
leucine rich repeat containing 43
There are no transcripts associated with this gene.

View Full Product Details