Product Details

SNP ID
rs61732427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50482954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCAGCCAGGGTGCTGTGCCCC[C/T]GTCCAGAGCTGCAGCTGAGCCCCAT
Phenotype
MIM: 608682 MIM: 606774 MIM: 603560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADM2 PubMed Links

Gene Details

Gene
ADM2
Gene Name
adrenomedullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253845.1 549 UTR 3 NP_001240774.1
XM_017028940.1 549 UTR 3 XP_016884429.1
Gene
MIOX
Gene Name
myo-inositol oxygenase
There are no transcripts associated with this gene.

Gene
SBF1
Gene Name
SET binding factor 1
There are no transcripts associated with this gene.

View Full Product Details