Product Details

SNP ID
rs3732707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160568858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGAGCATCGTGGGGAGCTGGC[C/T]GTTAAGGATGCCAATGCCAAGCTCT
Phenotype
MIM: 602970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KPNA4 PubMed Links

Gene Details

Gene
KPNA4
Gene Name
karyopherin subunit alpha 4
There are no transcripts associated with this gene.

Gene
KRT8P12
Gene Name
keratin 8 pseudogene 12
There are no transcripts associated with this gene.

View Full Product Details