Product Details

SNP ID
rs6787916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52833699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCGGGCCCCCTTCAGGTCCTC[C/G]TCCTTCACAGGGGGGCGCTTCTTCT
Phenotype
MIM: 600564
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITIH4 PubMed Links

Gene Details

Gene
ITIH4
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
There are no transcripts associated with this gene.

Gene
ITIH4-AS1
Gene Name
ITIH4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MUSTN1
Gene Name
musculoskeletal, embryonic nuclear protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205853.3 1061 Missense Mutation GAC,GAG D20E NP_995325.3
Gene
TMEM110
Gene Name
transmembrane protein 110
There are no transcripts associated with this gene.

Gene
TMEM110-MUSTN1
Gene Name
TMEM110-MUSTN1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198974.2 1061 Missense Mutation GAC,GAG D310E NP_001185903.2

View Full Product Details