Product Details

SNP ID
rs35870568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24676321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCTCCTGGAGGAGACCGAGGTG[C/T]GGGCCCAAGAAGGGCCCAGAAGAGT
Phenotype
MIM: 610922
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPAP1 PubMed Links

Gene Details

Gene
NPAP1
Gene Name
nuclear pore associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018958.2 928 Missense Mutation CGG,TGG R152W NP_061831.2

View Full Product Details