Product Details

SNP ID
rs571520861
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:155754165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGGTATTTTGTTTTTCCAATTC[C/T]AAAAGCTTTACTCACAGAATTCTCT
Phenotype
MIM: 610393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GON4L PubMed Links

Gene Details

Gene
GON4L
Gene Name
gon-4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282856.1 Intron NP_001269785.1
NM_001282858.1 Intron NP_001269787.1
NM_001282860.1 Intron NP_001269789.1
NM_001282861.1 Intron NP_001269790.1
NM_032292.5 Intron NP_115668.4
XM_005245284.3 Intron XP_005245341.1
XM_005245286.3 Intron XP_005245343.1
XM_006711393.3 Intron XP_006711456.1
XM_006711394.3 Intron XP_006711457.1
XM_011509658.2 Intron XP_011507960.1
XM_011509659.2 Intron XP_011507961.1
Gene
LOC100505728
Gene Name
uncharacterized LOC100505728
There are no transcripts associated with this gene.

Gene
MSTO2P
Gene Name
misato family member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details