Product Details

SNP ID
rs62001025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:147390853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACCGGAGGCGGCGGCGGCAGCA[A/G]CAGGGCCAGGTGGTAGCTCGGGGCT
Phenotype
MIM: 608948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZIC4 PubMed Links

Gene Details

Gene
ZIC4
Gene Name
Zic family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168378.1 Intron NP_001161850.1
NM_001168379.1 Intron NP_001161851.1
NM_001243256.1 Intron NP_001230185.1
NM_032153.5 Intron NP_115529.2
XM_017007314.1 Intron XP_016862803.1

View Full Product Details