Product Details
- SNP ID
-
rs62001038
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:108150081 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACTAGGCTGTCGTCACTATCTTC[C/T]TTTTTCACAGTCCTGTCTGAAGGAG
- Phenotype
-
MIM: 601581
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NRCAM
PubMed Links
Gene Details
- Gene
- NRCAM
- Gene Name
- neuronal cell adhesion molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037132.2 |
3903 |
Silent Mutation |
AAA,AAG |
K1248K |
NP_001032209.1 |
NM_001193582.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1155K |
NP_001180511.1 |
NM_001193583.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1136K |
NP_001180512.1 |
NM_001193584.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1124K |
NP_001180513.1 |
NM_005010.4 |
3903 |
Silent Mutation |
AAA,AAG |
K1127K |
NP_005001.3 |
XM_005250373.2 |
3903 |
Silent Mutation |
AAA,AAG |
K1248K |
XP_005250430.1 |
XM_005250383.2 |
3903 |
Silent Mutation |
AAA,AAG |
K1158K |
XP_005250440.1 |
XM_005250385.2 |
3903 |
Silent Mutation |
AAA,AAG |
K1143K |
XP_005250442.1 |
XM_006716003.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1252K |
XP_006716066.1 |
XM_006716007.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1240K |
XP_006716070.1 |
XM_006716012.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1159K |
XP_006716075.1 |
XM_006716014.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1147K |
XP_006716077.1 |
XM_011516253.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1255K |
XP_011514555.1 |
XM_011516255.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1255K |
XP_011514557.1 |
XM_011516256.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1251K |
XP_011514558.1 |
XM_011516257.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1249K |
XP_011514559.1 |
XM_011516258.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1249K |
XP_011514560.1 |
XM_011516259.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1245K |
XP_011514561.1 |
XM_011516261.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1239K |
XP_011514563.1 |
XM_011516262.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1236K |
XP_011514564.1 |
XM_011516265.2 |
3903 |
Intron |
|
|
XP_011514567.1 |
XM_011516266.2 |
3903 |
Intron |
|
|
XP_011514568.1 |
XM_011516267.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1162K |
XP_011514569.1 |
XM_011516268.2 |
3903 |
Silent Mutation |
AAA,AAG |
K1159K |
XP_011514570.1 |
XM_011516269.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1150K |
XP_011514571.1 |
XM_011516270.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1146K |
XP_011514572.1 |
XM_011516271.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1130K |
XP_011514573.1 |
XM_017012236.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1242K |
XP_016867725.1 |
XM_017012237.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1236K |
XP_016867726.1 |
XM_017012238.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1232K |
XP_016867727.1 |
XM_017012239.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1230K |
XP_016867728.1 |
XM_017012240.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1229K |
XP_016867729.1 |
XM_017012241.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1233K |
XP_016867730.1 |
XM_017012242.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1217K |
XP_016867731.1 |
XM_017012243.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1201K |
XP_016867732.1 |
XM_017012244.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1200K |
XP_016867733.1 |
XM_017012245.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1197K |
XP_016867734.1 |
XM_017012246.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1149K |
XP_016867735.1 |
XM_017012247.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1140K |
XP_016867736.1 |
XM_017012248.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1140K |
XP_016867737.1 |
XM_017012249.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1139K |
XP_016867738.1 |
XM_017012250.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1143K |
XP_016867739.1 |
XM_017012251.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1137K |
XP_016867740.1 |
XM_017012252.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1136K |
XP_016867741.1 |
XM_017012253.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1141K |
XP_016867742.1 |
XM_017012254.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1133K |
XP_016867743.1 |
XM_017012255.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1133K |
XP_016867744.1 |
XM_017012256.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1137K |
XP_016867745.1 |
XM_017012257.1 |
3903 |
Silent Mutation |
AAA,AAG |
K1136K |
XP_016867746.1 |
XM_017012258.1 |
3903 |
UTR 3 |
|
|
XP_016867747.1 |
XM_017012259.1 |
3903 |
UTR 3 |
|
|
XP_016867748.1 |
View Full Product Details