Product Details

SNP ID
rs375202254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94395261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGGAATTGATAAGAAACATGGA[A/T]ATTACTTTAAAAAATGAAAACATAA
Phenotype
MIM: 120160
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COL1A2 PubMed Links

Gene Details

Gene
COL1A2
Gene Name
collagen type I alpha 2 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000089.3 Intron NP_000080.2

View Full Product Details