Product Details

SNP ID
rs73232059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111999660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGCAGGGAAACCCTCCAGGGC[C/T]CTTTCTTTTAACGTAAGGCTGCGGG
Phenotype
MIM: 607953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD10 PubMed Links

Gene Details

Gene
ABHD10
Gene Name
abhydrolase domain containing 10
There are no transcripts associated with this gene.

Gene
TAGLN3
Gene Name
transgelin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008272.1 Intron NP_001008273.1
NM_001008273.1 Intron NP_001008274.1
NM_013259.2 Intron NP_037391.2

View Full Product Details