Product Details

SNP ID
rs35978996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111999599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCAGAAATGGTTAATGGACGG[C/G]ACGGTAAGGCCGGCAGCGATCTCGG
Phenotype
MIM: 607953
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABHD10 PubMed Links

Gene Details

Gene
ABHD10
Gene Name
abhydrolase domain containing 10
There are no transcripts associated with this gene.

Gene
TAGLN3
Gene Name
transgelin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008272.1 729 Silent Mutation GGC,GGG G59G NP_001008273.1
NM_001008273.1 729 Silent Mutation GGC,GGG G59G NP_001008274.1
NM_013259.2 729 Silent Mutation GGC,GGG G59G NP_037391.2

View Full Product Details