Product Details

SNP ID
rs532197891
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:156337977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAGGCCAGTTTGAGTTTGTGCA[A/G]AGAAGCGAACAGGTTAAGAGAAGAG
Phenotype
MIM: 600114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT3 PubMed Links

Gene Details

Gene
CCT3
Gene Name
chaperonin containing TCP1 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008800.2 Intron NP_001008800.1
NM_005998.4 Intron NP_005989.3
Gene
TSACC
Gene Name
TSSK6 activating cochaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304817.1 Intron NP_001291746.1
NM_001304818.1 Intron NP_001291747.1
NM_001304819.1 Intron NP_001291748.1
NM_001304820.1 Intron NP_001291749.1
NM_001304821.1 Intron NP_001291750.1
NM_001304822.1 Intron NP_001291751.1
NM_001304823.1 Intron NP_001291752.1
NM_001304824.1 Intron NP_001291753.1
NM_001304825.1 Intron NP_001291754.1
NM_001304826.1 Intron NP_001291755.1
NM_144627.4 Intron NP_653228.1

View Full Product Details