Product Details

SNP ID
rs59999845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42774039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTGTAGCTGACATTTTCCCC[C/T]GGCAAATGCAGTACCATACCCTCTT
Phenotype
MIM: 610907 MIM: 601844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPS25 PubMed Links

Gene Details

Gene
VPS25
Gene Name
vacuolar protein sorting 25 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032353.3 Intron NP_115729.1
XM_017025224.1 Intron XP_016880713.1
Gene
WNK4
Gene Name
WNK lysine deficient protein kinase 4
There are no transcripts associated with this gene.

View Full Product Details