Product Details

SNP ID
rs41281507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55523585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTTGTGTTTTGGTGAAAGACA[A/G]TGTCACTGTCATGTAATATGTAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP36 PubMed Links

Gene Details

Gene
CFAP36
Gene Name
cilia and flagella associated protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282761.1 Intron NP_001269690.1
NM_080667.6 Intron NP_542398.3
XM_011532499.1 Intron XP_011530801.1
XM_017003254.1 Intron XP_016858743.1
XM_017003255.1 Intron XP_016858744.1

View Full Product Details