Product Details

SNP ID
rs763433349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85780542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACAGCAGCGGCAGGCGCCTCCT[C/G]GGGGCTCTCCCTGCAGCCGTGCCCA
Phenotype
MIM: 604886
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC8 PubMed Links

Gene Details

Gene
EMC8
Gene Name
ER membrane protein complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142288.1 Intron NP_001135760.1
NM_006067.4 Intron NP_006058.1
XM_011522813.2 Intron XP_011521115.1
XM_017022867.1 Intron XP_016878356.1

View Full Product Details