Product Details

SNP ID
rs148525399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46702609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAACCTTCCAGGAGTTTCATCTT[A/T]AATTTGTCCCAGGAAAGTTTCATCT
Phenotype
MIM: 602732 MIM: 616454
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP1 PubMed Links

Gene Details

Gene
ARHGAP1
Gene Name
Rho GTPase activating protein 1
There are no transcripts associated with this gene.

Gene
ZNF408
Gene Name
zinc finger protein 408
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184751.1 Intron NP_001171680.1
NM_024741.2 Intron NP_079017.1

View Full Product Details