Product Details

SNP ID
rs73454037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46701011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTGGAGGAGCTGAGAAGAGGA[A/C]GCTCACTTCCGGCGTAGGGAGGCTT
Phenotype
MIM: 602732 MIM: 616454
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP1 PubMed Links

Gene Details

Gene
ARHGAP1
Gene Name
Rho GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004308.3 245 Intron NP_004299.1
XM_011520095.1 245 Intron XP_011518397.1
XM_017017720.1 245 Intron XP_016873209.1
Gene
ZNF408
Gene Name
zinc finger protein 408
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184751.1 245 Intron NP_001171680.1
NM_024741.2 245 UTR 5 NP_079017.1

View Full Product Details