Product Details
- SNP ID
-
rs35527067
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97930366 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTTCAGAATGAGGTGTTTCTTG[G/T]ATTGTTTTATGAGAGGGGGCTTTAG
- Phenotype
-
MIM: 610715
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HEMGN
PubMed Links
Gene Details
- Gene
- HEMGN
- Gene Name
- hemogen
- Gene
- TRMO
- Gene Name
- tRNA methyltransferase O
There are no transcripts associated with this gene.
View Full Product Details