Product Details

SNP ID
rs191398516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99985081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTCCAGGAATTCTAAAACCAGA[A/G]TCAAATAAAATGTAAACTGTTAAAA
Phenotype
MIM: 609170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERP44 PubMed Links

Gene Details

Gene
ERP44
Gene Name
endoplasmic reticulum protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015051.2 Intron NP_055866.1

View Full Product Details