Product Details

SNP ID
rs113834286
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:26322214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACATCTGGCTGCAGATGTGGCCC[A/G]ATTCACTGATGCTCCACGTCTGGCG
Phenotype
MIM: 114280 MIM: 609151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIM1L PubMed Links

Gene Details

Gene
AIM1L
Gene Name
absent in melanoma 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039775.3 4983 Missense Mutation TCG,TTG S1616L NP_001034864.2
XM_005245918.2 4983 Missense Mutation TCG,TTG S1616L XP_005245975.1
XM_011541672.1 4983 Missense Mutation TCG,TTG S1604L XP_011539974.1
XM_011541673.2 4983 Missense Mutation TCG,TTG S1673L XP_011539975.1
Gene
CD52
Gene Name
CD52 molecule
There are no transcripts associated with this gene.

Gene
UBXN11
Gene Name
UBX domain protein 11
There are no transcripts associated with this gene.

View Full Product Details