Product Details

SNP ID
rs561345749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122910498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTCTTCACCCTGCCCTCCTAC[A/C]CTGCCCTCTGCTTCTACAAGAAGGC
Phenotype
MIM: 609298
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEMA5B PubMed Links

Gene Details

Gene
SEMA5B
Gene Name
semaphorin 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031702.3 Intron NP_001026872.2
NM_001256346.1 Intron NP_001243275.1
NM_001256347.1 Intron NP_001243276.1
NM_001256348.1 Intron NP_001243277.1
XM_017006638.1 Intron XP_016862127.1
XM_017006639.1 Intron XP_016862128.1

View Full Product Details